Polyvinylpyrrolidone, M.W. K 30 (MW ∼ 40,000)

Supplier: TCI America

P0472-25G P0472-500G
TCP0472-025GEA 67.28 CAD
TCP0472-025G TCP0472-500G
Polyvinylpyrrolidone, M.W. K 30 (MW ∼ 40,000)
Polyvinylpyrrolidone

CAS Number: 9003-39-8
Form: Powder
Color: White


Product Categories: Materials Chemistry, Polymer Research Reagents

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR