N-Phthaloyl-L-glutamic anhydride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P0620-1G P0620-5G
TCP0620-001GEA 82.63 CAD
TCP0620-001G TCP0620-5G
N-Phthaloyl-L-glutamic anhydride ≥98.0% (by titrimetric analysis)
N-Phthaloyl-L-glutamic anhydride

CAS Number: 25830-77-7
MDL Number: MFCD00067324
Molecular Formula: C13H9NO5
Molecular Weight: 259.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 205
Specific rotation [a]20/D: -43 deg (C=3, Dioxane)

Formula: C₁₃H₉NO₅
Melting Pt: 205 °C
MDL Number: MFCD00067324
CAS Number: 25830-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4340425dd0d3044bfbc466096964554a685a2c2c&vid=4340425dd0d3044bfbc466096964554a685a2c2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR