Polyethylene glycol 4,000

Supplier: TCI America

P0885-500G P0885-25G
TCP0885-500GEA 78.67 CAD
TCP0885-500G TCP0885-25G
Polyethylene glycol 4,000
Polyethylene glycol 4,000

CAS Number: 25322-68-3
Form: Crystals
Color: White
Melting point (°C): 58
Flash Point (°C): 270


Product Categories: Bioscience, Drug Delivery Systems (DDS) Research Reagents, Materials Chemistry, Polymer Research Reagents

Formula: H(OCH₂CH₂)nOH
Boiling Pt: > 250 °C (1013 hPa)
Melting Pt: 53…58 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD01779612
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

Average molecular weight 2,600 to 3,800
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR