Propionic acid ≥99% (by GC), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

U330-07 U330-09
JTU330-7CS 2343.54 CAD
JTU330-7 JTU330-9
Propionic acid ≥99% (by GC), J.T.Baker®
Propionic acid
Formula: C₃H₆O₂
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: –21 °C
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00002756
CAS Number: 79-09-4
EINECS: 201-176-3
UN: 3463
ADR: 8 (3),II
REACH: 01-2119486971-24
Merck Index: 13,07917

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃CH₂COOH) (by GC) ≥99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR