Tris(2-ethylhexyl)phosphate ≥98.0%

Supplier: TCI America

Synonyms: Trioctyl phosphate, Phosphoric acid tris(2-ethylhexyl ester), Tris(2-ethylhexyl) phosphate

P1022-25ML P1022-500ML
TCP1022-025MLEA 39.5 CAD
TCP1022-025ML TCP1022-500ML
Tris(2-ethylhexyl)phosphate ≥98.0%
Tris(2-ethylhexyl) phosphate

CAS Number: 78-42-2
MDL Number: MFCD00009491
Molecular Formula: C24H51O4P
Molecular Weight: 434.64
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 215
Flash Point (°C): 204
Specific Gravity (20/20): 0.93


Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives, Resin Additives
Other Product Names / Synonyms: Phosphoric Acid Trioctyl Ester, Phosphoric Acid Tris(2-ethylhexyl) Ester, Trioctyl Phosphate

Formula: C₂₄H₅₁O₄P
MW: 434.64 g/mol
Boiling Pt: 215 °C
Density: 0.93 (20 °C)
Flash Pt: 204 °C
MDL Number: MFCD00009491
CAS Number: 78-42-2
EINECS: 201-116-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=267c04cdc182bcfaf0b72b124da3eb883e926b2a&vid=267c04cdc182bcfaf0b72b124da3eb883e926b2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR