Polyethyleneimine 50% in water, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: PolyaziridinePolyethyleniminePolyethylenimine (branched)Polyaziridine Mw 25000PEICorcatPolymin PEthyleneimine PolymerAziridinePoly(ethyleneimine)

Management of Change (MOC) category = N

BAKRU230-08 U230-08
CAJTU230-8EA 436.08 CAD
CAJTU230-8 JTU230-8
Polyethyleneimine 50% in water, J.T.Baker®
Polyethyleneimine
Formula: (CH₂CH₂NH)n
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance (colorless to light yellow) Passes Test
Identification (by IR) Passes Test
pH of 5% Solution in water, For Information Only
Residue after Evaporation, For Information Only
Viscosity, For Information Only

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR