Polyethylene glycol 400, liquid, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
PEG 400
Management of Change (MOC) category = N
BAKRU216-01
BAKRU216-07
BAKRU216-09
U216-09
CAJTU216-1EA
2434.94
CAD
CAJTU216-1
CAJTU216-7
CAJTU216-9
JTU216-9
Polyethylene glycol 400, liquid, J.T.Baker®
Polyethylene glycol 400
Formula:
H(OCH₂CH₂)nOH MW: 400 g/mol Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 4…8 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 245 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Appearance | Passes Test |
Identification (by IR) | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...