n-Pentane ≥98%, BAKER ANANYZED™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Pentane

Management of Change (MOC) category = N

BAKRT007-01 T007-09 T007-07
CAJTT007-1EA 14399.38 CAD
CAJTT007-1 CAJTT007-7 CAJTT007-9 JTT007-9 JTT007-7
n-Pentane ≥98%, BAKER ANANYZED™
n-Pentane

Lot analysis on label.

Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: -130
Density: 0.63 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃(CH₂)₃CH₃) (by GC, corrected for water) ≥98 %
Color (APHA) ≤10
Residue after Evaporation ≤0.0020 %
Sulfur Compounds (as S) ≤0.005 %
Water (by KF, coulometric) ≤0.02 %
Boiling Range (95%)Lower 34 - 36 °C
Boiling Range (95%)Upper 34 - 40 °C
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR