2,4,6-Trichlorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2,4,6-TBA
T1518-10G
T1518-25G
TCT1518-010GEA
85.32
CAD
TCT1518-010G
TCT1518-025G
2,4,6-Trichlorobenzoic acid ≥98.0% (by GC, titration analysis)
2,4,6-Trichlorobenzoic acid
CAS Number: 50-43-1
MDL Number: MFCD00060699
Molecular Formula: C7H3Cl3O2
Molecular Weight: 225.45
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 165
Formula:
C₇H₃Cl₃O₂ MW: 225.46 g/mol Melting Pt: 165 °C |
MDL Number:
MFCD00060699 CAS Number: 50-43-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...