Tetraethylene glycol monomethyl ether ≥98.0%

Supplier: TCI America

T1372-5G T1372-25G
TCT1372-005GEA 120.16 CAD
TCT1372-005G TCT1372-025G
Tetraethylene glycol monomethyl ether ≥98.0%
Tetraethylene glycol monomethyl ether

CAS Number: 23783-42-8
MDL Number: MFCD00041756
Molecular Formula: C9H20O5
Molecular Weight: 208.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 125
Specific Gravity (20/20): 1.07


Product Categories: Bioscience, PEGylation Reagents
Other Product Names / Synonyms: Methyl Tetraglycol, mPEG4-Alcohol

Formula: C₉H₂₀O₅
Boiling Pt: 125 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00041756
CAS Number: 23783-42-8
EINECS: 245-883-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3e418a76f884bc2df14e25c56bddcffe73149d7&vid=c3e418a76f884bc2df14e25c56bddcffe73149d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR