(3R)-(+)-3-(Trifluoroacetamido)pyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1369-1G T1369-25G
TCT1369-001GEA 322.32 CAD
TCT1369-001G TCT1369-25G
(3R)-(+)-3-(Trifluoroacetamido)pyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)
(3R)-(+)-3-(Trifluoroacetamido)pyrrolidine hydrochloride

CAS Number: 141043-16-5
MDL Number: MFCD02094762
Molecular Formula: C6H9F3N2O
Molecular Weight: 218.60
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 235
Specific rotation [a]20/D: 27 deg (C=1, H2O)

Formula: C₆H₉F₃N₂O
Melting Pt: 235 °C
MDL Number: MFCD02094762
CAS Number: 141043-16-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971927bbb205d8acfc60419883fdb0fa4bbc9814&vid=971927bbb205d8acfc60419883fdb0fa4bbc9814&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR