Thiazolidine-2-carboxylic acid ≥98.0%
Supplier: TCI America
Synonyms:
1,3-Thiazolidine-2-carboxylic acid, 2-Carboxy-1,3-thiazolidine, 1,3-Thiazolane-2-carboxylic acid
T1324-5G
T1324-1G
TCT1324-005GEA
83.74
CAD
TCT1324-005G
TCT1324-001G
Thiazolidine-2-carboxylic acid ≥98.0%
Thiazolidine-2-carboxylic acid
CAS Number: 16310-13-7
MDL Number: MFCD00068005
Molecular Formula: C4H7NO2S
Molecular Weight: 133.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 182
Other Product Names / Synonyms: Tetrahydrothiazole-2-carboxylic Acid
Formula:
C₄H₇NO₂S Melting Pt: 182 °C |
MDL Number:
MFCD00068005 CAS Number: 65126-70-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...