Paraffin Oil, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

BAKRS894-00 BAKRS894-07
CAJTS894-0CS 1979.74 CAD
CAJTS894-0 CAJTS894-7
Paraffin Oil, J.T.Baker®
Paraffin Oil
Boiling Pt: 300…500 °C (1013 hPa)
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 215 °C (closed cup)
CAS Number: 8012-95-1
EINECS: 232-384-2
Merck Index: 13,07093

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test
Product Information (not specifications): Saybolt Viscosity @ 100
degrees F (sec).... 345 - 355


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR