Tetrafluorophthalic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0986-25G T0986-5G
TCT0986-025GEA 382.63 CAD
TCT0986-025G TCT0986-005G
Tetrafluorophthalic acid ≥98.0% (by titrimetric analysis)
Tetrafluorophthalic acid

CAS Number: 652-03-9
MDL Number: MFCD00002407
Molecular Formula: C8H2F4O4
Molecular Weight: 238.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 154

Formula: C₈H₂F₄O₄
Boiling Pt: 341 °C (1013 hPa)
Melting Pt: 154 °C
MDL Number: MFCD00002407
CAS Number: 652-03-9
EINECS: 211-483-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca8ee828c16645ab143ba0ba5c173d6f227a25c8&vid=ca8ee828c16645ab143ba0ba5c173d6f227a25c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR