1,2,4,5-Tetramethylimidazole ≥98.0%

Supplier: TCI America

Synonyms: 1,2,4,5-Tetramethyl-1H-imidazole

T0971-25G T0971-100G
TCT0971-025GEA 458.23 CAD
TCT0971-025G 76220-492
1,2,4,5-Tetramethylimidazole ≥98.0%
1,2,4,5-Tetramethylimidazole

CAS Number: 1739-83-9
MDL Number: MFCD00059162
Molecular Formula: C7H12N2
Molecular Weight: 124.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White

Formula: C₇H₁₂N₂
MDL Number: MFCD00059162
CAS Number: 1739-83-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7a2945c65ca7475a8494a0b2e136e699c562425&vid=c7a2945c65ca7475a8494a0b2e136e699c562425&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR