Methyl tridecanoate ≥98.0% (by GC)

Supplier: TCI America

T0960-25ML T0960-100ML
TCT0960-025MLEA 371.82 CAD
TCT0960-025ML TCT0960-100ML
Methyl tridecanoate ≥98.0% (by GC)
Methyl tridecanoate

CAS Number: 1731-88-0
MDL Number: MFCD00008977
Molecular Formula: C14H28O2
Molecular Weight: 228.38
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 163
Melting point (°C): 7
Flash Point (°C): 113
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Tridecanoic Acid Methyl Ester

Formula: C₁₄H₂₈O₂
Boiling Pt: 163 °C
Melting Pt: 7 °C
Density: 0.87 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008977
CAS Number: 1731-88-0
EINECS: 217-054-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b61cb2868938f1a5aab5c8c6337708448ebf86e&vid=b8411cb41d4aac7a17cf2ba91307bf95c4436f18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR