Triethylene glycol diacetate ≥98.0%
Supplier: TCI America
Synonyms:
Triethylene Glycol Diacetate
T0923-25G
T0923-500G
TCT0923-025GEA
33.18
CAD
TCT0923-025G
TCT0923-500G
Triethylene glycol diacetate ≥98.0%
Triethylene glycol diacetate
CAS Number: 111-21-7
MDL Number: MFCD00026213
Molecular Formula: C10H18O6
Molecular Weight: 234.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 300
Flash Point (°C): 174
Specific Gravity (20/20): 1.12
Formula:
C₁₀H₁₈O₆ MW: 234.25 g/mol Boiling Pt: 300 °C Density: 1.12 (20 °C) Flash Pt: 174 °C |
MDL Number:
MFCD00026213 CAS Number: 111-21-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...