Triethylene glycol monobutyl ether ≥98.0% (by GC)
Supplier: TCI America
T0915-500ML
T0915-25ML
TCT0915-500MLEA
86.79
CAD
TCT0915-500ML
TCT0915-025ML
Triethylene glycol monobutyl ether ≥98.0% (by GC)
Triethylene glycol monobutyl ether
CAS Number: 143-22-6
MDL Number: MFCD00020606
Molecular Formula: C10H22O4
Molecular Weight: 206.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 278
Melting point (°C): -35
Flash Point (°C): 156
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: Butyl Triglycol
Formula:
C₁₀H₂₂O₄ Boiling Pt: 278 °C Melting Pt: –35 °C Density: 0.99 (20 °C) Flash Pt: 156 °C |
MDL Number:
MFCD00020606 CAS Number: 143-22-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...