Trityl alcohol ≥99.0%

Supplier: TCI America

Synonyms: Triphenylmethanol, Triphenylcarbinol

T0516-25G T0516-500G
TCT0516-025GEA 59.57 CAD
TCT0516-025G TCT0516-500G
Trityl alcohol ≥99.0%
Trityl alcohol

CAS Number: 76-84-6
MDL Number: MFCD00004445
Molecular Formula: C19H16O
Molecular Weight: 260.34
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 380
Melting point (°C): 163


Other Product Names / Synonyms: Trityl Alcohol

Formula: C₁₉H₁₆O
MW: 260.34 g/mol
Boiling Pt: 360
Melting Pt: 163 °C
Density: 1.1884 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004445
CAS Number: 76-84-6
EINECS: 200-988-5
Merck Index: 13,09810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bee50f3e3cc70047b0d6e366846c92361e253370&vid=bee50f3e3cc70047b0d6e366846c92361e253370&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR