n-Tridecane ≥99.0%
Supplier: TCI America
|
Danger
|
T0411-25ML
T0411-500ML
T0411-100ML
TCT0411-025MLEA
53.06
CAD
TCT0411-025ML
TCT0411-500ML
TCT0411-100ML
n-Tridecane ≥99.0%
n-Tridecane
CAS Number: 629-50-5
MDL Number: MFCD00008979
Molecular Formula: C13H28
Molecular Weight: 184.37
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 235
Melting point (°C): -5
Flash Point (°C): 79
Specific Gravity (20/20): 0.76
Formula:
H₃C(CH₂)₁₁CH₃ Boiling Pt: 235 °C Melting Pt: –5 °C Density: 0.76 (20 °C) Flash Pt: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008979 CAS Number: 629-50-5 EINECS: 211-093-4 UN: 3295 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...