Perchloric acid 60 - 62%, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

BAKR9656-00 BAKR9656-33
CAJT9656-0CS 2121.88 CAD
CAJT9656-0 CAJT9656-33
Perchloric acid 60 - 62%, BAKER ANALYZED® ACS
Perchloric acid

Lot analysis on label.

Formula: HClO₄
Boiling Pt: 160…181 °C (1013 hPa)
Melting Pt: –112.2 °C
Density: 1.53…1.768 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HClO₄) 60.0 - 62.0 %
ACS - Color (APHA) ≤10
ACS - Residue after Ignition ≤0.003 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Nitrogen Compounds (as N) ≤0.001 %
ACS - Sulfate (SO₄) ≤0.001 %
ACS - Silicate and Phosphate (as SiO₂) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR