Tribenzylamine ≥99.0% (by GC, titration analysis)

Supplier: TCI America

T0341-500G T0341-25G
TCT0341-500GEA 232.26 CAD
TCT0341-500G TCT0341-025G
Tribenzylamine ≥99.0% (by GC, titration analysis)
Tribenzylamine

CAS Number: 620-40-6
MDL Number: MFCD00004773
Molecular Formula: C21H21N
Molecular Weight: 287.41
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 390
Melting point (°C): 93
Flash Point (°C): 203

Formula: N(CH₂C₆H₅)₃
Boiling Pt: 390 °C
Melting Pt: 93 °C
Density: 0.9912 g/cm³ (95 °C)
Flash Pt: 203 °C
Storage Temperature: Ambient
MDL Number: MFCD00004773
CAS Number: 620-40-6
EINECS: 210-638-3
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=deb70096ec457166f1ef116ecf6d4559ad483141&vid=ee39970c5f1d526f07ef500848561336d208ae04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR