p-Toluenesulfonanilide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T0282-25G T0282-500G
TCT0282-025GEA 75.54 CAD
TCT0282-025G TCT0282-500G
p-Toluenesulfonanilide ≥98.0% (by HPLC, titration analysis)
p-Toluenesulfonanilide

CAS Number: 68-34-8
MDL Number: MFCD00025989
Molecular Formula: C13H13NO2S
Molecular Weight: 247.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 102

Formula: C₁₃H₁₃NO₂S
Melting Pt: 102 °C
MDL Number: MFCD00025989
CAS Number: 68-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88382da958db9631d5c86a5235aadfcba8a124c3&vid=88382da958db9631d5c86a5235aadfcba8a124c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR