Perchloric acid 69 - 72%, BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = N
BAKR9652-00
BAKR9652-01
BAKR9652-04
9652-33
CAJT9652-0CS
2206.26
CAD
CAJT9652-0
CAJT9652-1
CAJT9652-4
JT9652-33
Perchloric acid 69 - 72%, BAKER ANALYZED® ACS
Perchloric acid
Lot analysis on label.
Formula:
HClO₄ Boiling Pt: 160…181 °C (1013 hPa) Melting Pt: –112.2 °C Density: 1.53…1.768 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 EINECS: 231-512-4 UN: 1873 ADR: 5.1,I |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (HClO₄) | 69.0 - 72.0 % |
ACS - Color (APHA) | ≤ 10 |
ACS - Residue after Ignition | ≤ 0.003 % |
ACS - Chloride (Cl) | ≤ 0.001 % |
ACS - Nitrogen Compounds (as N) | ≤ 0.001 % |
ACS - Sulfate (SO₄) | ≤ 0.001 % |
ACS - Silicate and Phosphate (as SiO₂) | ≤ 5 ppm |
ACS - Heavy Metals (by ICP-OES) | ≤ 1 ppm |
Trace Impurities - Iron (Fe) | ≤ 1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...