2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers) ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2,2,4,4-Tetramethylcyclobutane-1,3-diol
T0145-25G
T0145-5G
TCT0145-025GEA
507.18
CAD
TCT0145-025G
TCT0145-5G
2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers) ≥98.0% (by GC)
2,2,4,4-Tetramethyl-1,3-cyclobutanediol (mixture of isomers)
CAS Number: 3010-96-6
MDL Number: MFCD00001329
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 127
Flash Point (°C): 51
Formula:
C₈H₁₆O₂ MW: 144.21 g/mol Melting Pt: 127 °C Flash Pt: 51 °C |
MDL Number:
MFCD00001329 CAS Number: 3010-96-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...