Sulfolane ≥99.0%

Supplier: TCI America
Warning

T0115-500G T0115-25G
TCT0115-500GEA 168.48 CAD
TCT0115-500G TCT0115-025G
Sulfolane ≥99.0%
Sulpholane

CAS Number: 126-33-0
MDL Number: MFCD00005484
Molecular Formula: C4H8O2S
Molecular Weight: 120.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 285
Flash Point (°C): 168
Freezing point (°C): 25


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Sulfolane, Tetramethylene Sulfone

Formula: C₄H₈O₂S
Boiling Pt: 285 °C
Flash Pt: 168 °C
MDL Number: MFCD00005484
CAS Number: 126-33-0
EINECS: 204-783-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6e60b09b93d9889d6df5d4e72edfa0a402d913f&vid=c6e60b09b93d9889d6df5d4e72edfa0a402d913f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR