1,2,3,4-Tetrahydroquinoline ≥95.0%

Supplier: TCI America

T0113-25ML T0113-500ML T0113-100ML
TCT0113-025MLEA 35.38 CAD
TCT0113-025ML TCT0113-500ML TCT0113-100ML
1,2,3,4-Tetrahydroquinoline ≥95.0%
1,2,3,4-Tetrahydroquinoline

CAS Number: 635-46-1
MDL Number: MFCD00006693
Molecular Formula: C9H11N
Molecular Weight: 133.19
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 251
Flash Point (°C): 113
Freezing point (°C): 13
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C

Formula: C₉H₁₁N
Boiling Pt: 251 °C
Density: 1.06 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00006693
CAS Number: 635-46-1
EINECS: 211-237-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=832cdada89b3cf8cff167e218bff97534a2868ad&vid=832cdada89b3cf8cff167e218bff97534a2868ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR