n-Tetradecane ≥99.0%

Supplier: TCI America
Danger

Synonyms: Tetradecane

T0079-25ML T0079-500ML
TCT0079-025MLEA 48.14 CAD
TCT0079-025ML TCT0079-500ML
n-Tetradecane ≥99.0%
n-Tetradecane

CAS Number: 629-59-4
MDL Number: MFCD00008986
Molecular Formula: C14H30
Molecular Weight: 198.39
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 253
Melting point (°C): 6
Flash Point (°C): 100
Specific Gravity (20/20): 0.76

Formula: H₃C(CH₂)₁₂CH₃
MW: 198.39 g/mol
Boiling Pt: 253 °C
Melting Pt: 6 °C
Density: 0.76 (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008986
CAS Number: 629-59-4
EINECS: 211-096-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed14fe66fa439f06579fe3d4632f6318aa316cbc&vid=7c7f28699b09d01b890be99628d285a2c0261a76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR