Terephthalaldehyde ≥98.0%

Supplier: TCI America

T0010-500G T0010-25G T0010-100G
TCT0010-500GEA 339.7 CAD
TCT0010-500G TCT0010-025G TCT0010-100G
Terephthalaldehyde ≥98.0%
Terephthalaldehyde

CAS Number: 623-27-8
MDL Number: MFCD00006949
Molecular Formula: C8H6O2
Molecular Weight: 134.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 115
Flash Point (°C): 76


Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction)
Other Product Names / Synonyms: Benzene-1,4-dicarboxaldehyde

Formula: C₈H₆O₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.381 g/cm³ (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00006949
CAS Number: 623-27-8
EINECS: 210-784-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0be1b1b077e795fb257d3d76a7f546d66a5bb4d4&vid=0be1b1b077e795fb257d3d76a7f546d66a5bb4d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR