Dibutyl-L-(+)-tartrate ≥98% (by GC)

Supplier: TCI America

T0005-500G T0005-25G
TCT0005-500GEA 466.1 CAD
TCT0005-500G TCT0005-025G
Dibutyl-L-(+)-tartrate ≥98% (by GC)
Dibutyl-L-(+)-tartrate

CAS Number: 87-92-3
MDL Number: MFCD00009443
Molecular Formula: C12H22O6
Molecular Weight: 262.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 160
Specific Gravity (20/20): 1.09
Specific rotation [a]20/D: 13 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-(+)-Tartaric Acid Dibutyl Ester

Formula: C₁₂H₂₂O₆
Density: 1.09 (20 °C)
Flash Pt: 160 °C
MDL Number: MFCD00009443
CAS Number: 15763-01-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19769f45d1219beb7535a422b3ff2f73e16a0ece&vid=19769f45d1219beb7535a422b3ff2f73e16a0ece&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR