Trimellitic acid tris(2-ethylhexyl) ester ≥97.0%

Supplier: TCI America

B0881-25ML B0881-500ML
TCB0881-025MLEA 56.3 CAD
TCB0881-025ML TCB0881-500ML
Trimellitic acid tris(2-ethylhexyl) ester ≥97.0%
Trimellitic acid tris(2-ethylhexyl) ester

CAS Number: 3319-31-1
MDL Number: MFCD00053799
Molecular Formula: C33H54O6
Molecular Weight: 546.79
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 414
Flash Point (°C): 256
Specific Gravity (20/20): 0.99


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: 1,2,4-Benzenetricarboxylic Acid Tris(2-ethylhexyl) Ester, Trimellitic Acid Tris(2-ethylhexyl) Ester, Trioctyl 1,2,4-Benzenetricarboxylate, Trioctyl Trimellitate

Formula: C₃₃H₅₄O₆
Boiling Pt: 414 °C
Density: 0.99 (20 °C)
Flash Pt: 256 °C
MDL Number: MFCD00053799
CAS Number: 3319-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c019069f8cfdec9733d4a4eab6e7f3f6fe817413&vid=c019069f8cfdec9733d4a4eab6e7f3f6fe817413&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR