Taurine 98,0

Supplier: TCI America

Synonyms: 2-Aminoethanesulphonic acid, 2-Aminoethanesulfonic acid

A0295-500G A0295-25G
TCA0295-500GEA 69.36 CAD
TCA0295-500G TCA0295-025G
Taurine 98,0
Taurine

CAS Number: 107-35-7
MDL Number: MFCD00008197
Molecular Formula: C2H7NO3S
Molecular Weight: 125.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 300


Other Product Names / Synonyms: 2-Aminoethylsulfonic Acid, Taurine

Formula: C₂H₇NO₃S
MW: 125.15 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00008197
CAS Number: 107-35-7
EINECS: 203-483-8
Merck Index: 13,09163
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc41fec3cabc37efab37e5367e4a776aa29e67b0&vid=dc41fec3cabc37efab37e5367e4a776aa29e67b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR