TCLP extraction fluid 2 for use in EPA 1311 toxicity characteristic leaching procedure pH 2.88 ± 0.05 (25°C)
Supplier: Ricca Chemical
|
Danger
|
R8378000-4A
R8378000-20F
RC83781EA
210.14
CAD
RC83781
RC83785
TCLP extraction fluid 2 for use in EPA 1311 toxicity characteristic leaching procedure pH 2.88 ± 0.05 (25°C)
TCLP extraction fluid 2
pH 2.88 ± 0.05 at 25°C.
Specification Test Results
Appearance | Clear, colorless, slight Acetic odor |
pH at 25 °C (traceable to NIST SRM 185 & 186) | 2.88 ± 0.02 pH at 25.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...