Tetrahydrofuran 99 GC, corrected for water content Stabilised, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

Stabilization: stab
M.O.C. Category N

MACR8498-04 MACR8498-08 MACR8498-19 MACR8498-26
CAMK849804CS 3176.62 CAD
CAMK849804 CAMK849808 CAMK849819 CAMK849826
Tetrahydrofuran 99 GC, corrected for water content Stabilised, AR® ACS, Macron Fine Chemicals™
Tetrahydrofuran
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: -108.5
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance (clear, colorless liquid) Passes Test
ACS - Assay (C₄H₈O) (by GC, corrected for water) ≥99.0 %
ACS - Color (APHA) ≤20
ACS - Peroxide (as H₂O₂)(at time of packaging) ≤0.015 %
ACS - Residue after Evaporation ≤0.03 %
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Contains 0.025% Butylated Hydroxytoluene as a stabilizer.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR