Trichloroacetic acid (TCA) 99,0 acidimetry on anhydrous substance, crystals, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
TCA
M.O.C. Category N
MACR2928-08
MACR2928-10
CAMK292808EA
218.94
CAD
CAMK292808
CAMK292810
Trichloroacetic acid (TCA) 99,0 acidimetry on anhydrous substance, crystals, AR® ACS, Macron Fine Chemicals™
Trichloroacetic acid (TCA)
Crystals.
Formula:
Cl₃CCOOH MW: 163.39 g/mol Boiling Pt: 196 Melting Pt: 54…56 °C Density: 1.63 g/cm³ (20 °C) Flash Pt: 113 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004177 CAS Number: 76-03-9 EINECS: 200-927-2 UN: 1839 ADR: 8 II Merck Index: 13,09700 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (Cl₃CCOOH) (dried basis)(by acidimetry) | ≥99.0 % |
ACS - Clarity of Solution | Passes Test |
ACS - Insoluble Matter | ≤0.01 % |
ACS - Residue after Ignition | ≤0.03 % |
ACS - Chloride (Cl) | ≤0.002 % |
ACS - Nitrate (NO₃) | ≤0.002 % |
ACS - Phosphate (PO₄) | ≤5 ppm |
ACS - Sulfate (SO₄) | ≤0.02 % |
ACS - Heavy Metals (as Pb) | ≤0.002 % |
ACS - Iron (Fe) | ≤0.001 % |
ACS - Substances Darkened by H₂SO₄ | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...