Tetrahydrofuran 99,8 no preservatives_contains, ChromAR® LC, UV, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

M.O.C. Category N

2858-08 MACR2858-06
MK285808CS 1933.43 CAD
MK285808 CAMK285806
Tetrahydrofuran 99,8 no preservatives_contains, ChromAR® LC, UV, Macron Fine Chemicals™
Tetrahydrofuran

ChromAR—Solvents that meet ACS specifications and are suitable for liquid chromatography and UV-spectrophotometry. 

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: -108.5
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₄H₈O) (by GC, corrected for water) ≥99.8 %
Peroxide (as H₂O₂) ≤0.015 %
Residue after Evaporation ≤5 ppm
Water (H₂O)(by KF, coulometric) ≤0.03 %
Optical Absorbance (1-cm path vs water) - 212 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 225 nm ≤0.50
Optical Absorbance (1-cm path vs water) - 250 nm ≤0.17
Optical Absorbance (1-cm path vs water) - 300 nm ≤0.01
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR