2,3,5-Triphenyltetrazolium chloride (TTC) 90-100
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC
M.O.C. Category N
BAKRX135-01
BAKRX135-03
CAJTX135-1EA
236.38
CAD
CAJTX135-1
CAJTX135-3
2,3,5-Triphenyltetrazolium chloride (TTC) 90-100
2,3,5-Triphenyltetrazolium chloride
Formula:
C₁₉H₁₅ClN₄ MW: 334.81 g/mol Boiling Pt: ∼360 °C (1013 hPa) Melting Pt: 235 Storage Temperature: Refrigerator |
MDL Number:
MFCD00011963 CAS Number: 298-96-4 EINECS: 206-071-6 Merck Index: 13,09815 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Identification (by IR) | Passes Test |
Loss on Drying at 105°C | ≤5.0 % |
Residue after Ignition | Passes Test |
Sensitivity | Passes Test |
Solubility | Passes Test |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...