Triethylamine ≥99% (by GC), BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: (Diethylamino)ethane, TEA

Management of Change (MOC) category = N

BAKRW635-07 BAKRW635-09 W635-07
CAJTW635-7CS 2926.93 CAD
CAJTW635-7 CAJTW635-9 JTW635-7
Triethylamine ≥99% (by GC), BAKER ANALYZED®, J.T.Baker®
Triethylamine

Lot analysis on label.

Formula: (C₂H₅)₃N
MW: 101.19 g/mol
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.729 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3,II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay ((C₂H₅)₃N) (by GC, corrected for water) ≥ 99 %
Color (APHA) ≤ 15
Water (H₂O) ≤ 1 %
Identification (by IR) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR