Tetrahydrofuran ≥99.0% stabilized, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Butylene oxideTHFDiethylene oxide

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Management of Change (MOC) category = N

BAKRV530-01 V530-01
CAJTV530-1EA 2460.72 CAD
CAJTV530-1 JTV530-1
Tetrahydrofuran ≥99.0% stabilized, J.T.Baker®
Tetrahydrofuran
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
MDL Number: MFCD00005356
CAS Number: 109-99-9
UN: 2056
ADR: 3,II
Merck Index: 12,09351
https://ca.vwr.com/stibo/web/std.lang.all/95/19/789519.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₄H₈O) (by GC, corrected for water) ≥99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR