, BAKER ANALYZED® for LC-MS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

BAKR9835-02 BAKR9835-03
CAJT9835-2CS 3650.83 CAD
CAJT9835-2 CAJT9835-3
, BAKER ANALYZED® for LC-MS, J.T.Baker®
Acetonitrile with 0.1% (v/v) trifluoroacetic acid

For use in liquid chromatography and mass spectrometry. Clear, colorless liquid.

Storage Temperature: Ambient
EINECS: 200-929-3
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Trifluoroacetic Acid 0.095 - 0.105 %
Gradient Elution Test (a.u.) - 254 nm ≤0.01
Residue after Evaporation ≤1.0 ppm
Water (H₂O) ≤200 ppm
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤50 ppb
Trace Impurities - Aluminum (Al) ≤50.0 ppb
Trace Impurities - * Calcium (Ca) ≤50.0 ppb
Trace Impurities - Iron (Fe) ≤50.0 ppb
Trace Impurities - Magnesium (Mg) ≤50.0 ppb
Trace Impurities - Potassium (K) ≤50 ppb
Trace Impurities - * Sodium (Na) ≤50.0 ppb
* May change over time due to extraction from glass container.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR