2,2,4-Trimethylpentane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

Management of Change (MOC) category = N

BAKR9478-01 BAKR9478-03
CAJT9478-1CS 3242.22 CAD
CAJT9478-1 CAJT9478-3
2,2,4-Trimethylpentane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS
2,2,4-Trimethylpentane

Lot analysis on label.

Formula: (CH₃)₂CHCH₂C(CH₃)₃
Boiling Pt: 99.2 °C (1013 hPa)
Melting Pt: –107 °C
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay ((CH₃)₃CCH₂CH(CH₃)₂) (by GC, corrected for water) ≥99.0 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Water-Soluble Titrable Acid, meq/g ≤0.0003
Sulfur Compounds (as S) ≤0.005 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR