Triethanolamine (trolamine) ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

BAKR9468-01 BAKR9468-03 9468-03
CAJT9468-7EA 6462.91 CAD
CAJT9468-7 CAJT9468-1 CAJT9468-3 JT9468-3
Triethanolamine (trolamine) ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Triethanolamine (Trolamine)

Lot analysis on label.

Formula: C₆H₁₅NO₃
Boiling Pt: 335.4 °C (1013 hPa)
Melting Pt: 21.6 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 179 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (by GC) ≥99.0 %
Residue after Ignition ≤0.005 %
Water (by KF, volumetric) ≤0.2 %
Trace Impurities - Iron (Fe) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR