Chemware® Teflon® FEP Gas Sampling Bags, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Life Sciences
D1075002-10 D1075001-10 D1075003-10 D1075005-10 D1075007-10
32310-300CS 1629.8 CAD
32310-300 32310-301 32310-302 32310-304 32310-306
Chemware® Teflon® FEP Gas Sampling Bags, Saint-Gobain Performance Plastics
Bags Gas Bags
For gas sample analysis, sampling smokestack exhaust systems, monitoring air quality, and calibrating instruments in the field.
  • Inert to almost all chemicals
  • Low permeability limits sample loss
  • Pressure less, and fill is rated by volume
Made from 5 mil thick FEP fluoropolymer film, these flexible, sturdy bags are heat-sealed on all sides for complete chemical resistance and leakproof operation. Available with either a nickel-plated brass on/off valve which accepts ¹/₄" Int.Ø. flexible tubing or PTFE/silicone septum in polypropylene housing, for use with a hypodermic needle.
Packaging: Individually packaged.