Toluene ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

BAKR9460-07 BAKR9460-R BAKR9460-22 BAKR9460-03 BAKR9460-33 BAKR9460-05
CAJT9460-7EA 982.13 CAD
CAJT9460-7 CAJT9460-R CAJT9460-11 CAJT9460-22 CAJT9460-3 CAJT9460-33 CAJT9460-5
Toluene ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Toluene

Lot analysis on label.

Formula: C₆H₅CH₃
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₆H₅CH₃) (by GC) ≥99.5 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Substances Darkened by H₂SO₄ Passes Test
Sulfur Compounds (as S) ≤0.003 %
Water (by KF, coulometric) ≤0.03 %
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR