Tetrahydrofuran ≥99.5% (by GC, corrected for water content) stabilized, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 100 - 400 ppm
Management of Change (MOC) category = N

BAKR9450-R BAKR9450-01 BAKR9450-03 BAKR9450-33 BAKR9450-05
CAJT9450-REA 15415.62 CAD
CAJT9450-R CAJT9450-1 CAJT9450-3 CAJT9450-33 CAJT9450-5
Tetrahydrofuran ≥99.5% (by GC, corrected for water content) stabilized, BAKER ANALYZED® ACS, J.T.Baker®
Tetrahydrofuran

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Formula: C₄H₈O
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₄H₈O) (by GC, corrected for water) ≥99.5 %
Color (APHA) ≤20
Peroxide (as H₂O₂) ≤0.015 %
Residue after Evaporation ≤0.0300 %
Water (by KF, coulometric) ≤0.05 %
Specific Gravity at 25°/25°C 0.884 - 0.886
Stabilizer (BHT) 100 - 400 ppm
Acidity Passes Test
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR