Tetrahydrofuran 99,8 GC, corrected for water content no preservatives_contains, BAKER ANALYZED® HPLC HPLC, UV, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

M.O.C. Category N

9441-02 9441-03
CAJT9441-33CS 3088.09 CAD
CAJT9441-33 JT9441-2 JT9441-3
Tetrahydrofuran 99,8 GC, corrected for water content no preservatives_contains, BAKER ANALYZED® HPLC HPLC, UV, J.T.Baker®
Tetrahydrofuran

BAKER ANALYZED™ HPLC Reagent—High purity solvents for use in analytical and preparative separations. All are optimized for all of your Liquid Chromatography applications, including HPLC, UHPLC, gel permeation chromatography and UV-spectrophotometric analysis.


BAKER ANALYZED™ HPLC Reagent—High purity solvents for use in analytical and preparative separations. All are optimized for all of your Liquid Chromatography applications, including HPLC, UHPLC, gel permeation chromatography and UV-spectrophotometric analysis.

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: -108.5
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-320 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.030
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.15
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.45
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤212
Assay (by GC, corrected for water) ≥99.8 %
Peroxide (as H2O2) (atTime of Fill) ≤0.005 %
Residue after Evaporation ≤2.0 ppm
Water (by KF, coulometric) ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR