VWR® Traceable® Flip-Stick™ Thermometer
Supplier: VWR International
15551-002 37000-422
15551-002EA 111.23 CAD
15551-002 37000-422
VWR® Traceable® Flip-Stick™ Thermometer
Thermometers Digital Thermometers
A stainless steel fold-up protected probe permits carrying on the wrist-strap or in a pocket.
  • Traceable®
  • Flip-to-open design
  • Shockproof
  • Reads in °C and °F
Thermometer is shockproof in drop tests up to 1.5m (5'). Other features include °C/°F switch, minimum and maximum memory readings over any time period, and a hold switch (freezes display to capture a temperature). Unit has a big-digit, 1.9 cm (3/4") display.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.