n-Pentane ≥99.0% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Pentane

Management of Change (MOC) category = NI

9333-03 9333-03
JT9333-3CS 3215.08 CAD
JT9333-3 CAJT9333-3
n-Pentane ≥99.0% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
n-Pentane
Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: –130 °C
Density: 0.63 g/cm³ (20 °C)
Flash Pt: –35 °C
MDL Number: MFCD00009498
CAS Number: 109-66-0
UN: 1265
ADR: 3,II
Merck Index: 13,07193
https://ca.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL) ≤5
Assay (CH₃(CH₂)₃CH₃) (by GC, corrected for water) ≥99.0 %
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR