n-Pentane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Pentane

Management of Change (MOC) category = N

9331-03 9331-33
CAJT9331-3CS 311.14 CAD
CAJT9331-3 CAJT9331-33 JT9331-3 JT9331-33 CAJT9331-68
n-Pentane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
n-Pentane

BAKER ANALYZED™ HPLC Reagent—High purity solvents for use in analytical and preparative separations. All are optimized for all of your Liquid Chromatography applications, including HPLC, UHPLC, gel permeation chromatography and UV-spectrophotometric analy

Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: -130
Density: 0.63 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-245 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤0.4
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤190
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.5 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.5 ppb
Assay (CH₃(CH₂)₃CH₃) (by GC, corrected for water) ≥99.0 %
Residue after Evaporation ≤1.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR