Potassium hydroxide 0.1 N in methanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Potash caustic, Caustic potash
Management of Change (MOC) category = N
BAKR5650-02
BAKR5650-03
CAJT5650-2CS
921.54
CAD
CAJT5650-2
CAJT5650-3
Potassium hydroxide 0.1 N in methanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Potassium hydroxide
Standardized at 25°C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to +/- 1 part per 1000, provided on label.
Formula:
KOH MW: 56.11 g/mol Density: 0.80 g/cm³ Flash Pt: 12.00 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 2924 ADR: 3 (8),II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Normality | 0.0995 - 0.1005 |
Chloride (Cl) | ≤10 ppm |
Nitrogen Compounds (as N) | ≤1 ppm |
ACS - Heavy Metals (as Pb) | ≤1 ppm |
Trace Impurities - Iron (Fe) | ≤0.5 ppm |
Appearance | Passes Test |
Standardization at 25°C traceable to NIST Standard Reference Material. | |
SRM No… |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...